Learn More
Haztech Systems, Inc Hazcat Test Tube Clamp - Brass plated Steel 5"
SDP
Catalog No. NC9249853
Encompass
Hazcat's test tube lamp is perfect for holding the 13x100 mm test tube when heating unknown substances. 5" Brass plated steel.
Catalog No. NC9249853 Supplier Haztech Systems, Inc Supplier No. HW4014
Hazcat's test tube lamp is perfect for holding the 13x100 mm test tube when heating unknown substances. 5" Brass plated steel.