Learn More
Azzota Corp Test tube cover SV800
Catalog No. 5000887041
Encompass_Preferred
Test tube cover Used for long test tube on spectrophotometer SV800 Height 3 inch Diameter 2 inch
Catalog No. 50-008-87041 Supplier Azzota Corp Supplier No. S128
Test tube cover Used for long test tube on spectrophotometer SV800 Height 3 inch Diameter 2 inch