Learn More
Waters Corp Test Tube Domino, 14 mL Round Bottom, 24-Tube Capacity
Catalog No. NC3807264 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Test Tube Domino is a machine-readable consumable holder designed for the Andrew+ pipetting robot. It creates an interface between various consumable types and the robot, enabling seamless automation of laboratory workflows.
  • Holds up to 24 tubes
  • Equipped with QR codes for critical labware information
  • Equipped with ARUCO codes for spatial positioning
  • Designed to hold 14 mL round bottom test tubes
  • Indispensable for Andrew+ automation configurations
Catalog No. NC3807264 Supplier Waters Corp Supplier No. 186010165
The Test Tube Domino is a machine-readable consumable holder designed for the Andrew+ pipetting robot. It creates an interface between various consumable types and the robot, enabling seamless automation of laboratory workflows.
  • Holds up to 24 tubes
  • Equipped with QR codes for critical labware information
  • Equipped with ARUCO codes for spatial positioning
  • Designed to hold 14 mL round bottom test tubes
  • Indispensable for Andrew+ automation configurations