Learn More
Electron Microscopy Sciences Test Tubes Flint, 16 mm O.D., 125 mm height, Orange Label, Orange Label
SDP
Catalog No. 5025521
Encompass_Preferred
Flint glass, round bottom, plain top, handy fused-in color patches accept any type of ink or pencil.
O.D: 16 mm
Height: 125 mm
Label: Orange
Catalog No. 50-255-21 Supplier Electron Microscopy Sciences Supplier No. 64638O
Flint glass, round bottom, plain top, handy fused-in color patches accept any type of ink or pencil.
O.D: 16 mm
Height: 125 mm
Label: Orange