Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Test Tube Holder
SCI_ED

Catalog No. S72040 Shop All OHAUS Products
Change view
Click to view available options
Holds:
6.4 to 11.4 cm Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
S72040 6.4 to 11.4 cm Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72040 Supplier OHAUS™ Supplier No. 30400219

missing translation for 'validMainframeMsg'

Hands free mixing designed to fit on the Heavy Duty Vortex Mixer. Easily attached to the top of mixer and is magnetically secured.

  • Use with Heavy Duty Vortex Mixer
TRUSTED_SUSTAINABILITY

Specifications

Holds 6.4 to 11.4 cm Tubes
Tube Size 19 mm dia. x 6.4 to 11.4 cm L
Material Stainless Steel
Description Single Tube Holder, VXHD Vortex Mixers
For Use With (Equipment) Heavy Duty Vortex Mixer
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.