Learn More
Chemglass Life Sciences Test Tube Tong, 6" OAL
SDP
Catalog No. 501311228
Encompass_Preferred
Test Tube Tong, 6" OAL. Tongs are made of zinc coated steel, 6" overall length. Spring loaded jaw will hold a tube up to 1" in diameter.
Catalog No. 50-131-1228 Supplier Chemglass Life Sciences Supplier No. CG4018100
Test Tube Tong, 6" OAL. Tongs are made of zinc coated steel, 6" overall length. Spring loaded jaw will hold a tube up to 1" in diameter.