Learn More
Research Products International Corp TEST TUBE PEG RACK FOR 10-13MM TUBES, WHITE, 2/PKG
SDP
Catalog No. 50998197
Encompass
Hold sample collection tubes, reaction tubes and test tubes with Test Tube Peg Racks. Suitable for drying glass plates used in electrophoresis and chromatography plates. Will not float in a water bath. Chemically resistant and autoclavable.
Catalog No. 50-998-197 Supplier Research Products International Corp Supplier No. 59770013
Hold sample collection tubes, reaction tubes and test tubes with Test Tube Peg Racks. Suitable for drying glass plates used in electrophoresis and chromatography plates. Will not float in a water bath. Chemically resistant and autoclavable.