Learn More
Globe Scientific Test Tube Peg Racks
Catalog No. 21402989
Change view
Click to view available options
Positions:
50
96
Color:
Blue
Red
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions Color
21-402-989 96 Blue
21-402-990 96 Red
21-402-991 96 White
21-402-992 50 Blue
21-402-993 50 Red
21-402-994 50 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21-402-989 Supplier Globe Scientific Supplier No. 456150B
Only null left
Add to Cart
Add to Cart
Manufactured from durable reinforced polypropylene designed to allow easy viewing of tube contents
  • Fully Autoclavable
  • Manufactured from durable reinforced polypropylene (PP)
  • Peg design allows easy viewing for tube contents
  • Can be used to dry inverted tubes while minimizing collection or airborne contaminants
  • Available in various Color with 13 mm and 96 place
Specifications
Autoclavable Autoclavable
Color Blue
Holds Tubes (13 mm)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Product Type Test Tube Peg Rack
Shelves None
Array 96 Place