Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences OneRack Test Tube Rack 16 place for 16 mm tubes, Blue, 127 x 250 x 70 mm
SDP

Supplier:  Electron Microscopy Sciences 6161701

Encompass_Preferred

These durable racks are molded in a single, continuous piece - no assembly required and no detachable pieces. Racks are made of Delrin fiberglassreinforced polyoxymethylene, an engineered polymer developed by DuPont to have exceptional chemical and heat resistance. Molded index coordinates and flat labeling areas keep samples organized. Delrin is a registered trademark of E.I. du Pont de Nemours and Company. Temp. Range: -40 degree C to 140 degree C
Use: 16 mm tubes
Color: Blue
Size: 127 x 250 x 70 mm

Catalog No. 50-190-2676


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.