Learn More
Electron Microscopy Sciences OneRack Test Tube Rack 9 place for 30 mm tubes, Blue, 110 x 282 x 85 mm
SDP
Catalog No. 501902684
Encompass_Preferred
These durable racks are molded in a single, continuous piece - no assembly required and no detachable pieces. Racks are made of Delrin fiberglassreinforced polyoxymethylene, an engineered polymer developed by DuPont to have exceptional chemical and heat resistance. Molded index coordinates and flat labeling areas keep samples organized. Delrin is a registered trademark of E.I. du Pont de Nemours and Company. Temp. Range: -40 degree C to 140 degree C
Use: 30 mm tubes
Color: Blue
Size: 110 x 282 x 85 mm
Catalog No. 50-190-2684 Supplier Electron Microscopy Sciences Supplier No. 6161901
These durable racks are molded in a single, continuous piece - no assembly required and no detachable pieces. Racks are made of Delrin fiberglassreinforced polyoxymethylene, an engineered polymer developed by DuPont to have exceptional chemical and heat resistance. Molded index coordinates and flat labeling areas keep samples organized. Delrin is a registered trademark of E.I. du Pont de Nemours and Company. Temp. Range: -40 degree C to 140 degree C
Use: 30 mm tubes
Color: Blue
Size: 110 x 282 x 85 mm