Learn More
Cole-Parmer Cole-Parmer Cube test tube rack, PP, 1/ea
Catalog No. NC1438230 Shop All Cole-Parmer Products
Encompass_Preferred
Polypropylene cube rack features a total of 88 spaces on its four sides. All four sides can be used interchagealby for 12-mm through 25-mm diameter tubes. Autoclavable.
Catalog No. NC1438230 Supplier Cole-Parmer Supplier No. 0673937
Polypropylene cube rack features a total of 88 spaces on its four sides. All four sides can be used interchagealby for 12-mm through 25-mm diameter tubes. Autoclavable.