Learn More
Thomas Scientific Test Tube rack for 15 x 50 ml tubes
SDP
Catalog No. 501458112
Encompass_Preferred
Hinged lid (included) also removable Quik-CAL™ user calibration function Seamless, stainless steel chamber myBath 4L Includes racks for 1.5/2.0, 15 & 50ml tubes The myBath series is designed to guarantee superior accuracy and performance in a compact footprint. With four sizes to choose from, there is a myBath to fit every lab. Easy to use controls allow the user to digitaly select and monitor settings. Adjustable in 0.1ºC increments, the temperature is shown on a large, 3-digit display. The proprietary Quik-CAL™ feature allows for simple, in-lab calibration.* High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. All myBath models feature seamless, stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination. The larger 8L and 12L units have drains for hassle-free water changes. The 4L, 8L, and 12L models includ
Catalog No. 50-145-8112 Supplier Thomas Scientific Supplier No. 1200G04
Hinged lid (included) also removable Quik-CAL™ user calibration function Seamless, stainless steel chamber myBath 4L Includes racks for 1.5/2.0, 15 & 50ml tubes The myBath series is designed to guarantee superior accuracy and performance in a compact footprint. With four sizes to choose from, there is a myBath to fit every lab. Easy to use controls allow the user to digitaly select and monitor settings. Adjustable in 0.1ºC increments, the temperature is shown on a large, 3-digit display. The proprietary Quik-CAL™ feature allows for simple, in-lab calibration.* High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. All myBath models feature seamless, stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination. The larger 8L and 12L units have drains for hassle-free water changes. The 4L, 8L, and 12L models includ