Learn More
Bellco Glass, Inc. Test Tube Rack for 30mm Tubes
SDP
Catalog No. 502330689
Encompass_Preferred
Test Tube Racks are easy to assemble and available for use with common test tube sizesLightweight, stackable and autoclavableRacks fit 13 mm, 17 mm or 30 mm test tubesAll racks measure 246 mm x 104 mm x 64 mmRacks are submersible in water and resistant to common organic chemicals found in laboratories such as ethanol, NBF (neutral buffered formain) and xylene. Style-For 30 mm Tubes, Color-White, Height (mm)-64, Length (mm)-246, Width (mm)-104, Material-Polypropylene (PP), Carton Qty-1
Catalog No. 50-233-0689 Supplier Bellco Glass, Inc. Supplier No. 130000361
Test Tube Racks are easy to assemble and available for use with common test tube sizesLightweight, stackable and autoclavableRacks fit 13 mm, 17 mm or 30 mm test tubesAll racks measure 246 mm x 104 mm x 64 mmRacks are submersible in water and resistant to common organic chemicals found in laboratories such as ethanol, NBF (neutral buffered formain) and xylene. Style-For 30 mm Tubes, Color-White, Height (mm)-64, Length (mm)-246, Width (mm)-104, Material-Polypropylene (PP), Carton Qty-1