Learn More
Glas Col LLC Rotator Accessory
SDP
Catalog No. NC9840286
Encompass_Preferred
Holds a complete rack of stoppered test tubes for rotating or end over end also holds bottles, erlenmeyer flasks and other stoppered containers.For use with rotator, 099A RD4512 or 099A RD5512.
Catalog No. NC9840286 Supplier Glas Col LLC Supplier No. 099A RD50
Holds a complete rack of stoppered test tubes for rotating or end over end also holds bottles, erlenmeyer flasks and other stoppered containers.For use with rotator, 099A RD4512 or 099A RD5512.