Learn More
Cole-Parmer Cole-Parmer Test Tube Rack, PP 24 places
Supplier:  Cole-Parmer 0673917
Encompass_Preferred
Made from a special blend of polypropylene that sinks in a water bath and maintains stability even in agitated water. This double-tiered rack holds up to twenty-four 25-mm test tubes and is easy to stack when empty. Top tier features an alphanumeric grid for reference. Autoclavable.
Catalog No. NC1616610