Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Test Tube Racks, Biohazard Warning

Catalog No. 1479334E Shop All Fisherbrand Products
Change view
Click to view available options
Positions:
40
72
Holds:
Tubes (13 mm)
Tubes (20 to 25 mm)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions Holds
14-793-34E 40 Tubes (20 to 25 mm)
14-793-34A 72 Tubes (13 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-793-34E

missing translation for 'validMainframeMsg'

Easily identified in the lab and sterilization area by bright orange epoxy coating.

  • Designed for use with biologically hazardous materials
  • Also applicable in research with viruses, cancerous cells and recombinant DNA
  • Sterilization can be performed with tubes in racks prior to discarding hazardous materials
  • Universal biohazard symbol tag attached to each rack
  • Epoxy coated steel wire; chemical- and corrosion-resistant
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges

Specifications

Autoclavable Autoclavable
Color Orange
Holds Tubes (20 to 25 mm)
Material Steel
Positions 40
Shape Rectangular
Stackable No
Sterility Non-sterile
Diameter (Metric) Opening 25 mm
Product Type Test Tube Rack
Shelves None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.