Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Test Tube Stands

Catalog No. 21402969
Change view
Click to view available options
Holds:
Tubes (13 mm)
Tubes (16 mm)
Tubes (20 mm)
Tubes (25 mm)
Tubes (32 mm)
Color:
Blue
Red
White
Yellow
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Color
21-402-969 Tubes (13 mm) Blue
21-402-970 Tubes (13 mm) Red
21-402-971 Tubes (13 mm) White
21-402-972 Tubes (13 mm) Yellow
21-402-973 Tubes (16 mm) Blue
21-402-974 Tubes (16 mm) Red
21-402-975 Tubes (16 mm) White
21-402-976 Tubes (16 mm) Yellow
21-402-977 Tubes (20 mm) Blue
21-402-978 Tubes (20 mm) Red
21-402-979 Tubes (20 mm) White
21-402-980 Tubes (20 mm) Yellow
21-402-981 Tubes (25 mm) Blue
21-402-982 Tubes (25 mm) Red
21-402-983 Tubes (25 mm) White
21-402-984 Tubes (25 mm) Yellow
21-402-985 Tubes (32 mm) Blue
21-402-986 Tubes (32 mm) Red
21-402-987 Tubes (32 mm) White
21-402-988 Tubes (32 mm) Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 21-402-969 Supplier Globe Scientific Supplier No. 456113B
Add to Cart
Add to Cart

Manufactured from durable reinforced polypropylene, offers a three teired design allowing to hold tubes securely

  • Fully Autoclavable
  • Manufactured from durable reinforced polypropylene (PP)
  • Three tiered design holds tubes securely
  • Sturdy handles facilitate easy transport
  • 4 color options for organization of samples or benchtop

Specifications

Autoclavable Autoclavable
Color Blue
Holds Tubes (13 mm)
Material Polypropylene
Positions 31
Shape Rectangular
Stackable No
Sterility Non-sterile
Product Type Test Tube Holder
Shelves None
Array 31 Place
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.