Learn More
Heidolph™ Test Tube Trays
SDP
Catalog No. 13889381 Shop All Heidolph North America Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
13-889-381 Small
13-889-816 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-889-381 Supplier Heidolph™ Supplier No. 036303170
Add to Cart
Add to Cart
Heidolph™ Test Tube Trays for use with Reax Top/Reax Control Vibrating Shakers
  • Large Tray holds tubes and flasks up to 50 mL
  • Small Tray holds 20 mm diameter test tubes
  • Made from PTFE and Stainless Steel
Specifications
Product Type Test Tube Attachment
Material PTFE, Stainless Steel
For Use With (Application) Mixing Eppendorf Tubes, Vials and, Vessels; Ideal for Product Dissolution
For Use With (Equipment) Reax Top/Reax Control Vibrating Shakers
Size Small
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov