Learn More
Electron Microscopy Sciences Test Tubes, 12 mm O.D., 75 mm height, Blue Label, Blue Label
SDP
Catalog No. 5025513
Encompass_Preferred
These disposable glass tubes made of borosilicate glass have a round bottom, plain top, and labeling area. The handy fused-in color patches accept any type of ink or pencil.
O.D: 12 mm
Height: 75 mm
Label: Blue
Catalog No. 50-255-13 Supplier Electron Microscopy Sciences Supplier No. 64635B
These disposable glass tubes made of borosilicate glass have a round bottom, plain top, and labeling area. The handy fused-in color patches accept any type of ink or pencil.
O.D: 12 mm
Height: 75 mm
Label: Blue