Learn More
Electron Microscopy Sciences Test Tubes, 13 mm O.D., 100 mm height, Green Label, Green Label
SDP
Catalog No. 5025519
Encompass_Preferred
These disposable glass tubes made of borosilicate glass have a round bottom, plain top, and labeling area. The handy fused-in color patches accept any type of ink or pencil.
O.D: 13 mm
Height: 100 mm
Label: Green
Catalog No. 50-255-19 Supplier Electron Microscopy Sciences Supplier No. 64636G
These disposable glass tubes made of borosilicate glass have a round bottom, plain top, and labeling area. The handy fused-in color patches accept any type of ink or pencil.
O.D: 13 mm
Height: 100 mm
Label: Green