Learn More
BioVendor, LLC ENDOTOXIN FREE ENDOGRADE GLASS TEST TUBES
SDP
Catalog No. NC1140761
Encompass
TypeEndotoxin-free EndoGrade Glass TestTubeDescriptionHyglos' endotoxin-free glass test tubes are applied to dilute or aliquot samples and are ideal vessels for the dilution of the endotoxin standard contained in the EndoLISA kit and for sample collection after EndoTrap endotoxin removal.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore at 15–25°C.
Catalog No. NC1140761 Supplier BioVendor, LLC Supplier No. 800050
TypeEndotoxin-free EndoGrade Glass TestTubeDescriptionHyglos' endotoxin-free glass test tubes are applied to dilute or aliquot samples and are ideal vessels for the dilution of the endotoxin standard contained in the EndoLISA kit and for sample collection after EndoTrap endotoxin removal.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore at 15–25°C.