Learn More
Bellco Glass, Inc. Test Tubes without RIM 12*75 Borosilicate 3.3 ISO 4142 5ml
SDP
Catalog No. 502549560
Encompass_Preferred
Borosil Culture Test Tubes without Rim, ISO/DIN 4142, 5 mL, CS/800Borosil Test Tubes comply with IS 2618, ISO/ DIN 4142 and are designed with a round bottom. The test tubes are made thick-walled with borosilicate glass to be mechanically resistant. These test tubes have a high thermal shock resistance. Test tubes are used to hold, mix, and heat chemical experiments.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 5mLApprox O.D. x Length: 12mm x 75mmClass/ Quality Grade: Type IQuantity Per Case: 800
Catalog No. 50-254-9560 Supplier Bellco Glass, Inc. Supplier No. 452801202
Borosil Culture Test Tubes without Rim, ISO/DIN 4142, 5 mL, CS/800Borosil Test Tubes comply with IS 2618, ISO/ DIN 4142 and are designed with a round bottom. The test tubes are made thick-walled with borosilicate glass to be mechanically resistant. These test tubes have a high thermal shock resistance. Test tubes are used to hold, mix, and heat chemical experiments.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 5mLApprox O.D. x Length: 12mm x 75mmClass/ Quality Grade: Type IQuantity Per Case: 800