Learn More
Foxx Life Sciences Borosil® Reusable Test Tubes with Plain End, 38mL (25 x 100mm), 200/CS
Catalog No. 501720232
Encompass_Preferred
Borosil® Reusable Test Tubes w/ Plain End are constructed from mechanically strong, low-extractable borosilicate glass. The flame-polished, beaded rim confers strength, preventing chipping & cracking. Thick, uniform walls, excellent heat resistance, & rounded bottoms mean tubes can be reused without issue. Borosil® Reusable w/ Plain End Test Tubes comply w/ ISO/DIN 4142. Available in a variety of sizes from 1mL (7mm x 60 mm) to 170mL (38mm x 200mm). In collaboration w/ Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0232 Supplier Foxx Life Sciences Supplier No. 9820U07
Borosil® Reusable Test Tubes w/ Plain End are constructed from mechanically strong, low-extractable borosilicate glass. The flame-polished, beaded rim confers strength, preventing chipping & cracking. Thick, uniform walls, excellent heat resistance, & rounded bottoms mean tubes can be reused without issue. Borosil® Reusable w/ Plain End Test Tubes comply w/ ISO/DIN 4142. Available in a variety of sizes from 1mL (7mm x 60 mm) to 170mL (38mm x 200mm). In collaboration w/ Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.