Learn More
Bellco Glass, Inc. Test Tubes without RIM 7*60 Borosilicate 3.3 ISO 4142 1ml
SDP
Supplier:  Bellco Glass, Inc. 452801213
Encompass_Preferred
Borosil Culture Test Tubes without Rim, ISO/DIN 4142, 1 mL, CS/400Borosil Test Tubes comply with IS 2618, ISO/ DIN 4142 and are designed with a round bottom. The test tubes are made thick-walled with borosilicate glass to be mechanically resistant. These test tubes have a high thermal shock resistance. Test tubes are used to hold, mix, and heat chemical experiments.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 1mLApprox O.D. x Length: 7mm x 60mmClass/ Quality Grade: Type IQuantity Per Case: 400
Catalog No. 50-254-9549