Learn More
Sensonics Incorporated SITT-ROM
SDP
Catalog No. NC2092864
Encompass_Preferred
ROMANIAN UPSIT- 40 ITEM SMELL IDENTIFICATION TEST - MOQ 7 OR MORE TESTS - SINGLE USE - THE SIT ADMINISTRATION MANUAL WITH SCORE KEY (SITM-MAN) AND ADDITIONAL SCORE KEYS (SITK-KEY) ARE ORDERED SEPARATELY
This item is not returnable. View return policy
Catalog No. NC2092864 Supplier Sensonics Incorporated Supplier No. SITTROM
ROMANIAN UPSIT- 40 ITEM SMELL IDENTIFICATION TEST - MOQ 7 OR MORE TESTS - SINGLE USE - THE SIT ADMINISTRATION MANUAL WITH SCORE KEY (SITM-MAN) AND ADDITIONAL SCORE KEYS (SITK-KEY) ARE ORDERED SEPARATELY