Learn More
Sensonics Incorporated SITT-ESPA
SDP
Catalog No. NC2135492
Encompass_Preferred
SPANISH EUROPEAN (SPAIN) UPSIT- 40 ITEM SMELL IDENTIFICATION TEST - MOQ 7 OR MORE TESTS - SINGLE USE - THE SIT ADMINISTRATION MANUAL WITH SCORE KEY (SITM-MAN) AND ADDITIONAL SCORE KEYS (SITK-KEY) ARE ORDERED SEPARATELY
This item is not returnable. View return policy
Catalog No. NC2135492 Supplier Sensonics Incorporated Supplier No. SITTESPA
SPANISH EUROPEAN (SPAIN) UPSIT- 40 ITEM SMELL IDENTIFICATION TEST - MOQ 7 OR MORE TESTS - SINGLE USE - THE SIT ADMINISTRATION MANUAL WITH SCORE KEY (SITM-MAN) AND ADDITIONAL SCORE KEYS (SITK-KEY) ARE ORDERED SEPARATELY