Learn More
NEXPRING HEALTH TEST YOKE BUFFER W/GEN PK/20
Catalog No. NC0820816
Encompass
Refrigeration Medium - Test Yolk Buffer (TYB) with Gentamicin. Refrigeration Medium-TYB is for short term storage of human sperm at 2 deg. to 5 deg. C. 20x5ml.
Catalog No. NC0820816 Supplier NEXPRING HEALTH Supplier No. 9012920X5ML
Refrigeration Medium - Test Yolk Buffer (TYB) with Gentamicin. Refrigeration Medium-TYB is for short term storage of human sperm at 2 deg. to 5 deg. C. 20x5ml.