Learn More
US Biomax Inc Testis seminoma with testis tissue array, including pathology grade, TNM and clinical stage, 48 cases/48 cores, replacing TE481
SDP
Catalog No. NC3733353 Shop All US Biomax Inc Products
Encompass
Testis seminoma with testis tissue array, including pathology grade, TNM and clinical stage, 48 cases/48 cores, replacing TE481
Catalog No. NC3733353 Supplier US Biomax Inc Supplier No. TE481A
Testis seminoma with testis tissue array, including pathology grade, TNM and clinical stage, 48 cases/48 cores, replacing TE481