Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Testosterone Double Antibody RIA Kit

Catalog No. MP07189105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Tubes
500 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07189105 500 Tubes
MP07189102 100 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07189105 Supplier MP Biomedical™ Supplier No. 07189105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantitative Liquid Phase Radioimmunoassay

The Testosterone Double Antibody RIA Kit is to be used for detection and measurement of the total unconjugated form of this steroid. It does not distinguish between protein (albumin and SHBG) bound testosterone and true free testosterone which must be measured after a dialysis procedure.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
For Use With (Application) Endocrinology
Quantity 500 Tubes
Regulatory Status FDA Cleared (K791999)
Sample Volume 50 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.