Learn More
Arbor Assays Testosterone ELISA Kit, (5 Strip Plate)
SDP
Supplier:  Arbor Assays K080H5
Encompass_Preferred
The DetectX Testosterone ELISA Kits measure testosterone without extraction in serum and plasma, as well as in urine and dried fecal extracts.
Catalog No. 50-227-8522