Learn More
Enzo Life Sciences Testosterone ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900065
Encompass_Preferred
The Testosterone ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies. Sensitivity: 5.67 pg/ml (range 7.81 - 2,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, Steroid displacement reagent, pNpp Substrate, Stop solution
Catalog No. 50-658-123