Learn More
Drg International Inc DRG Testosterone ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA1559
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of testosterone in serum and plasma. In women, high levels of testosterone are generally found in hirsutism and virilization, polycystic ovaries, ovarian tumors, adrenal tumors and adrenal hyperplasia. In men, high levels of testosterone are associated to the hypothalamic pituitary unit diseases, testicular tumors, congenital adrenal hyperplasia and prostate cancer. Low levels of testosterone can be found in patients with the following diseases: Hypopituitarism, Klinefelter's syndrome, Testicular feminization, Orchidectomy and Cryptorchidism, enzymatic defects and some autoimmune diseases. The DRG Testosterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding.
Catalog No. 50-187-1236