Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Testosterone High Sensitivity Competitive ELISA Kit

Catalog No. EIATES
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIATES 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIATES Supplier Invitrogen™ Supplier No. EIATES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The multispecies Testosterone ELISA with improved sensitivity quantitates testosterone in serum, plasma, dried fecal samples, or urine. Principle of the method: The Competitive ELISA research-use-only kit is designed to quantitatively measure testosterone independent of species. A testosterone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with donkey anti-sheep IgG antibodies. A monoclonal sheep antibodies and testosterone-peroxidase conjugate is added to the wells. The testosterone conjugate and any testosterone in the sample will compete to bind to the sheep testosterone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound testosteron-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of testosterone present in the sample. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.8-1000 pg/mL
Assay Sensitivity 2.97 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chromone
Sample Type Fecal Extract, Plasma, Serum, Urine
Target Species Chemical
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; TestosteronAndroderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron;
Interassay CV <11%
Intraassay CV <6%
Kit Contents Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers, Dissociation Reagent, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone, Testosterone
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Urine 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Testosterone
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.