Learn More
Alpco Ltd Mouse / Rat Testosterone ELISA
SDP
Catalog No. 507513640
Encompass_Preferred
The Mouse / Rat Testosterone ELISA is a competitive immunoassay for the quantitative measurement of testosterone in rat and mouse serum or plasma.
  • Includes ready to use standards
  • Requires  10 μl of sample per well
  • Incubation time of only 1.5 hours
  • See Rat Control Set 55-DEV99RC for kit controls
For research use only. Not for use in diagnostic procedures.
Catalog No. 50-751-3640 Supplier Alpco Ltd Supplier No. 55TESMSE01
The Mouse / Rat Testosterone ELISA is a competitive immunoassay for the quantitative measurement of testosterone in rat and mouse serum or plasma.
  • Includes ready to use standards
  • Requires  10 μl of sample per well
  • Incubation time of only 1.5 hours
  • See Rat Control Set 55-DEV99RC for kit controls
For research use only. Not for use in diagnostic procedures.