Learn More
Cusabio Technology LLC Rat Testosterone,T ELISA Kit
Catalog No. NC2952848
Encompass_Preferred
This Rat Testosterone (T) ELISA Kit was designed for the quantitative measurement of Rat Testosterone (T) protein in serum, plasma, cell culture supernates, tissue homogenates. It is a Competitive ELISA kit, its detection range is 0.13 ng/mL-25.6 ng/mL and the sensitivity is 0.06 ng/mL. One kit contains 96 wells.
Catalog No. NC2952848 Supplier Cusabio Technology LLC Supplier No. CSBE05100R
This Rat Testosterone (T) ELISA Kit was designed for the quantitative measurement of Rat Testosterone (T) protein in serum, plasma, cell culture supernates, tissue homogenates. It is a Competitive ELISA kit, its detection range is 0.13 ng/mL-25.6 ng/mL and the sensitivity is 0.06 ng/mL. One kit contains 96 wells.