Learn More
Biomatik Corporation Testosterone (Testo) CLIA Kit
Supplier:  Biomatik Corporation EKU08623
Encompass_Preferred
Testosterone (Testo) CLIA Kit for detecting Testo in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 117.2-30,000pg/ml. Sensitivity: 40.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: T; Testesteron; Testeronun. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8651