Learn More
Electron Microscopy Sciences Testrite Print Roller, 4" wide
SDP
Catalog No. 5030506
Encompass_Preferred
These rollers are lightweight and sturdy, offering fine smooth action, yet they have ample strength to efficiently remove surplus water from prints and distribute an even pressure.
Width: 4"
Catalog No. 50-305-06 Supplier Electron Microscopy Sciences Supplier No. 7484404
These rollers are lightweight and sturdy, offering fine smooth action, yet they have ample strength to efficiently remove surplus water from prints and distribute an even pressure.
Width: 4"