Learn More
Medchemexpress LLC Tet-213 TFA | 99.9% | 1704.92 | 10 MG
SDP
Catalog No. 5000396965
Encompass_Preferred
Tet-213 TFA is an antimicrobial peptide with broad-spectrum antibacterial activity, which can promote infected wound repair.
  • Broad-spectrum antibacterial activity
  • Promotes infected wound repair
  • Shows significant antibacterial activity on 80% (16 of 20 strains) of *S. aureus* clinical isolates
  • Strongly reduces the growth and biofilm formation of *S. aureus*
Catalog No. 50-003-96965 Supplier Medchemexpress LLC Supplier No. HYP3459A10MG
Tet-213 TFA is an antimicrobial peptide with broad-spectrum antibacterial activity, which can promote infected wound repair.
  • Broad-spectrum antibacterial activity
  • Promotes infected wound repair
  • Shows significant antibacterial activity on 80% (16 of 20 strains) of *S. aureus* clinical isolates
  • Strongly reduces the growth and biofilm formation of *S. aureus*