Learn More
AnaSpec TET 830 modified/T-helper epitope from tetanus toxoid[AQYIKANSKFIGITEL] | MW 1797.2 | (1 mg)
Supplier:  AnaSpec AS60622
Encompass
TET 830 modified/T-helper epitope from tetanus toxoid[AQYIKANSKFIGITEL] ; MW 1797.2; (1 mg) TET 830 modified/T-helper epitope from tetanus toxoid is a universal human tetanus toxin T cell epitope. It induces T-cell activation and is used as a helper peptide in vaccinations.
Catalog No. NC0921367