Learn More
CAPTIVATE BIO LLC TET-Free FBS Premium
Catalog No. 5000876347
Encompass_Preferred
Captivate Bio s TET-Free FBS (aka TET-System Approved FBS or TET-approved) is tested to ensure the absence of detecting trace-level tetracycline and doxycycline residues that may interfere with gene expression and cellular function making this serum ideal for sensitive applications
Catalog No. 50-008-76347 Supplier CAPTIVATE BIO LLC Supplier No. FBS162TF
Captivate Bio s TET-Free FBS (aka TET-System Approved FBS or TET-approved) is tested to ensure the absence of detecting trace-level tetracycline and doxycycline residues that may interfere with gene expression and cellular function making this serum ideal for sensitive applications