Learn More
Takara Bio Tet System Approved FBS, US-Sourced, 50 ml
SDP
Catalog No. NC9082563
Encompass
The Tet-On Advanced Inducible Gene Expression System is a powerful, tightly regulated, tetracycline-inducible mammalian expression system that is controlled by the addition of doxycycline to the culture medium. The system includes a vector set, a control cell line and Tet System Approved FBS. 50ml
Catalog No. NC9082563 Supplier Takara Bio Supplier No. 631105
The Tet-On Advanced Inducible Gene Expression System is a powerful, tightly regulated, tetracycline-inducible mammalian expression system that is controlled by the addition of doxycycline to the culture medium. The system includes a vector set, a control cell line and Tet System Approved FBS. 50ml