Learn More
Takara Bio Tet System Approved FBS, 3 x 500 mL
SDP
Catalog No. NC0658188
Encompass
High-quality Tet System Approved Fetal Bovine Serum has been functionally tested to ensure that it permits the full range of tetracycline-regulated induction. This FBS has been collected from non-US sources, and approved for import and export by the USDA. 3 x 500 mL
Catalog No. NC0658188 Supplier Takara Bio Supplier No. 631367
High-quality Tet System Approved Fetal Bovine Serum has been functionally tested to ensure that it permits the full range of tetracycline-regulated induction. This FBS has been collected from non-US sources, and approved for import and export by the USDA. 3 x 500 mL