Learn More
New England Biolabs, Inc. TET2 - 400 units
SDP
Catalog No. 5000332619
Encompass
TET2 (Tet methylcytosine dioxygenase 2) is a demethylase and ten-eleven translocation protein that in the presence of α-ketoglutarate (αKG) and Fe(II)-dependent, oxidizes 5-Methylcytosine (5mC) in three consecutive steps to 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC), and 5-carboxycytosine (5caC), producing succinate (Suc) and CO2 as a byproduct.
  • Stored at -20°C
  • Specifically oxidizes 5mC, to 5hmC, 5fC, and 5caC in a stepwise manner
  • Supplied with reaction buffer, supplement, Fe(II) and DTT
Catalog No. 50-003-32619 Supplier New England Biolabs, Inc. Supplier No. M1524S
TET2 (Tet methylcytosine dioxygenase 2) is a demethylase and ten-eleven translocation protein that in the presence of α-ketoglutarate (αKG) and Fe(II)-dependent, oxidizes 5-Methylcytosine (5mC) in three consecutive steps to 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC), and 5-carboxycytosine (5caC), producing succinate (Suc) and CO2 as a byproduct.
  • Stored at -20°C
  • Specifically oxidizes 5mC, to 5hmC, 5fC, and 5caC in a stepwise manner
  • Supplied with reaction buffer, supplement, Fe(II) and DTT