Learn More
Genetex Inc Anti-TET3 antibody [C3], C-term, 25 μl
SDP
Supplier:  Genetex Inc GTX121453
Encompass
Host: Rabbit; The Anti-TET3 antibody [C3], C-term from GeneTex is a Rabbit Polyclonal antibody to TET3. The Anti-TET3 antibody [C3], C-term Recognizes Human, Mouse, Xenopus laevis, Bovine. The Anti-TET3 antibody [C3], C-term has been validated for the following Applications: ICC/IF, IHC-P, IP, WB, IHC-Wm, ChIP assay.
Catalog No. NC1561537