Learn More
List Biological Laboratories Inc Tetanus Toxin C-Fragment from Clostridium tetani, 10 μg
SDP
Catalog No. 501870954
Encompass_Preferred
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development project! Tetanus toxin is a potent neurotoxin (150 kDa) produced by the anaerobic bacterium Clostridium tetani. Intact toxin consists of two polypeptide chains connected through an interchain disulfide bond. The larger polypeptide (~100 kDa) consists of the N-terminal portion of the heavy chain, which contains the pH dependent translocation domain. The smaller polypeptide (~50 kDa), known as the C-fragment, consists of the binding portion and retains internalization properties. It has the advantage of being non-toxic and virtually devoid of any action on the nerve processes making it extremely useful for tracing fiber connections in the CNS. Tetanus toxin C-fragment is manufactured by enzyme digestion of native tetanus toxin produced in C. tetani. Each lot of C-fragment is tested for binding activity to GT1b ganglioside.
Catalog No. 50-187-0954 Supplier List Biological Laboratories Inc Supplier No. 193
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development project! Tetanus toxin is a potent neurotoxin (150 kDa) produced by the anaerobic bacterium Clostridium tetani. Intact toxin consists of two polypeptide chains connected through an interchain disulfide bond. The larger polypeptide (~100 kDa) consists of the N-terminal portion of the heavy chain, which contains the pH dependent translocation domain. The smaller polypeptide (~50 kDa), known as the C-fragment, consists of the binding portion and retains internalization properties. It has the advantage of being non-toxic and virtually devoid of any action on the nerve processes making it extremely useful for tracing fiber connections in the CNS. Tetanus toxin C-fragment is manufactured by enzyme digestion of native tetanus toxin produced in C. tetani. Each lot of C-fragment is tested for binding activity to GT1b ganglioside.