Learn More
List Biological Laboratories Inc Tetanus Toxin from Clostridium tetani, 25 μg
SDP
Catalog No. NC2941179
Encompass_Preferred
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development project! Tetanus toxin is a potent neurotoxin (150,000 Da) produced by the anaerobic bacterium Clostridium tetani. Intact toxin derived from culture supernatants consists of two polypeptide chains connected through an interchain disulfide bond. List Labs offers the native tetanus toxin produced from Clostridium tetani.
Catalog No. NC2941179 Supplier List Biological Laboratories Inc Supplier No. 190A
Add to Cart
Add to Cart
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development project! Tetanus toxin is a potent neurotoxin (150,000 Da) produced by the anaerobic bacterium Clostridium tetani. Intact toxin derived from culture supernatants consists of two polypeptide chains connected through an interchain disulfide bond. List Labs offers the native tetanus toxin produced from Clostridium tetani.