Learn More
Medchemexpress LLC Tetanus Toxin (830-844) | 126779-13-3 | 95.7% | 1724.99 | 10 MG
SDP
Catalog No. 5000399104
Encompass_Preferred
Tetanus Toxin (830-844) is a biologically active peptide derived from tetanus toxin, functioning as a TT830-844 CD4+ T-cell epitope. It is used in research for its ability to bind to a wide range of HLA-DRB molecules and activate CD4+ T-cell responses in a large part of the human population.
  • Promiscuous CD4+ T-cell epitope.
  • Binds to a wide range of HLA-DRB molecules.
  • Activates CD4+ T-cell responses in a large part of the human population.
Catalog No. 50-003-99104 Supplier Medchemexpress LLC Supplier No. HYP5425A10MG
Add to Cart
Add to Cart
Tetanus Toxin (830-844) is a biologically active peptide derived from tetanus toxin, functioning as a TT830-844 CD4+ T-cell epitope. It is used in research for its ability to bind to a wide range of HLA-DRB molecules and activate CD4+ T-cell responses in a large part of the human population.
  • Promiscuous CD4+ T-cell epitope.
  • Binds to a wide range of HLA-DRB molecules.
  • Activates CD4+ T-cell responses in a large part of the human population.