Learn More
Medchemexpress LLC Tetanus toxin (830-844) | 126779-13-3 | 95.7% | 1724.99 | 25 MG
SDP
Catalog No. 5000398681
Encompass_Preferred
Tetanus Toxin (830-844) is a biologically active peptide derived from tetanus toxin, specifically a TT830-844 CD4+ T-cell epitope. This peptide can bind to a wide range of HLA-DRB molecules, activating CD4+ T-cell responses in a significant portion of the human population. This product is for research use only.
  • Biologically active peptide
  • Tetanus toxin-derived peptide
  • CD4+ T-cell epitope
  • Binds to a wide range of HLA-DRB molecules
  • Activates CD4+ T-cell responses
  • For research use only
Catalog No. 50-003-98681 Supplier Medchemexpress LLC Supplier No. 25MGHYP5425A
Tetanus Toxin (830-844) is a biologically active peptide derived from tetanus toxin, specifically a TT830-844 CD4+ T-cell epitope. This peptide can bind to a wide range of HLA-DRB molecules, activating CD4+ T-cell responses in a significant portion of the human population. This product is for research use only.
  • Biologically active peptide
  • Tetanus toxin-derived peptide
  • CD4+ T-cell epitope
  • Binds to a wide range of HLA-DRB molecules
  • Activates CD4+ T-cell responses
  • For research use only