Learn More
Medchemexpress LLC Tetanus toxin (830-844) TFA | 95.67% | 1724.99 | 5 MG
SDP
Catalog No. 5000399105
Encompass_Preferred
Tetanus Toxin (830-844) TFA is a biologically active peptide that serves as a tetanus toxin-derived peptide TT830-844 CD4+ T-cell epitope. This promiscuous CD4+ T-cell epitope can bind to a wide range of HLA-DRB molecules, activating CD4+ T-cell responses in a large part of the human population.
  • Biologically active peptide
  • Tetanus toxin-derived peptide TT830-844 CD4+ T-cell epitope
  • Binds to a wide range of HLA-DRB molecules
  • Activates CD4+ T-cell responses
  • For research use only
Catalog No. 50-003-99105 Supplier Medchemexpress LLC Supplier No. HYP5425A5MG
Tetanus Toxin (830-844) TFA is a biologically active peptide that serves as a tetanus toxin-derived peptide TT830-844 CD4+ T-cell epitope. This promiscuous CD4+ T-cell epitope can bind to a wide range of HLA-DRB molecules, activating CD4+ T-cell responses in a large part of the human population.
  • Biologically active peptide
  • Tetanus toxin-derived peptide TT830-844 CD4+ T-cell epitope
  • Binds to a wide range of HLA-DRB molecules
  • Activates CD4+ T-cell responses
  • For research use only