Learn More
Drg International Inc DRG Tetanus Toxin IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3514R
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of IgG-class antibodies to Tetanus toxin in human serum or plasma. This product is for Research Use Only. The DRG Tetanus toxin IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Tetanus toxoid antigen. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7588